Product Details

SNP ID
rs11576266
Assay Type
Functionally tested
NCBI dbSNP Submissions
39
Location
Chr.1:156602367 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCCAGTCTAGCCTGGGAAACTTGGG[A/G]CCTTGAGTGGGATTAGATGGAGGAC
Phenotype
MIM: 608862
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GPATCH4 PubMed Links

Gene Details

Gene
GPATCH4
Gene Name
G-patch domain containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015590.3 Intron NP_056405.2
NM_182679.2 Intron NP_872620.1
XM_005245287.3 Intron XP_005245344.1
Gene
HAPLN2
Gene Name
hyaluronan and proteoglycan link protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021817.2 Intron NP_068589.1
XM_005245415.2 Intron XP_005245472.1
XM_011509853.2 Intron XP_011508155.1
XM_017002020.1 Intron XP_016857509.1
XM_017002021.1 Intron XP_016857510.1
Gene
NAXE
Gene Name
NAD(P)HX epimerase
There are no transcripts associated with this gene.

View Full Product Details