Product Details

SNP ID
rs12090418
Assay Type
Functionally tested
NCBI dbSNP Submissions
7
Location
Chr.1:112915594 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGAACTGGAGACTTCAAGGAGACT[A/G]AAGAACAGCTAGAACTGATACAGGT
Phenotype
MIM: 600682
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AKR7A2P1 PubMed Links

Gene Details

Gene
AKR7A2P1
Gene Name
aldo-keto reductase family 7 member A2 pseudogene 1
There are no transcripts associated with this gene.

Gene
SLC16A1
Gene Name
solute carrier family 16 member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166496.1 Intron NP_001159968.1
NM_003051.3 Intron NP_003042.3
XM_011542026.2 Intron XP_011540328.1
XM_011542027.2 Intron XP_011540329.1

View Full Product Details