Product Details

SNP ID
rs12324717
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:42554355 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TATTTTAACTGTCTTAAAATGTACA[A/C]TTCAGTGGCATTTACTATACTCAAT
Phenotype
MIM: 613429
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
HAUS2 PubMed Links

Gene Details

Gene
HAUS2
Gene Name
HAUS augmin like complex subunit 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130447.1 Intron NP_001123919.1
NM_001323629.1 Intron NP_001310558.1
NM_001323630.1 Intron NP_001310559.1
NM_001323631.1 Intron NP_001310560.1
NM_001323632.1 Intron NP_001310561.1
NM_018097.2 Intron NP_060567.1
XM_017022396.1 Intron XP_016877885.1
XM_017022397.1 Intron XP_016877886.1
Gene
LRRC57
Gene Name
leucine rich repeat containing 57
There are no transcripts associated with this gene.

View Full Product Details