Product Details

SNP ID
rs12062953
Assay Type
Functionally tested
NCBI dbSNP Submissions
40
Location
Chr.1:9949867 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCATTACTAAGATTTCTATTTTCCT[C/T]CCTCTTTTTCAGCACTTATTTCTAT
Phenotype
MIM: 610458 MIM: 608700
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LZIC PubMed Links

Gene Details

Gene
LZIC
Gene Name
leucine zipper and CTNNBIP1 domain containing
There are no transcripts associated with this gene.

Gene
NMNAT1
Gene Name
nicotinamide nucleotide adenylyltransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297778.1 Intron NP_001284707.1
NM_001297779.1 Intron NP_001284708.1
NM_022787.3 Intron NP_073624.2
XM_011541971.2 Intron XP_011540273.1
XM_017002107.1 Intron XP_016857596.1
XM_017002108.1 Intron XP_016857597.1

View Full Product Details