Product Details

SNP ID
rs12851982
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:71304918 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTCCACCCTGACCCCAATGCCTTT[G/T]ATTTCCTCATTCTCTCTTTCTTCTG
Phenotype
MIM: 300332 MIM: 300084
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ITGB1BP2 PubMed Links

Gene Details

Gene
ITGB1BP2
Gene Name
integrin subunit beta 1 binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303277.1 Intron NP_001290206.1
NM_012278.2 Intron NP_036410.1
XM_017029405.1 Intron XP_016884894.1
Gene
NONO
Gene Name
non-POU domain containing, octamer-binding
There are no transcripts associated with this gene.

View Full Product Details