Product Details

SNP ID
rs12743770
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:18113431 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGTCACCTCTGCAGCCCAAAAGA[A/G]CAGGAGAGACAGGTCGTGGGCAGGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IGSF21 PubMed Links

Gene Details

Gene
IGSF21
Gene Name
immunoglobin superfamily member 21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032880.4 Intron NP_116269.3
XM_011542319.2 Intron XP_011540621.1
XM_017002604.1 Intron XP_016858093.1
XM_017002605.1 Intron XP_016858094.1

View Full Product Details