Product Details

SNP ID
rs11573166
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:19977572 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTTTAGGGCAAAACCTAAATACAG[C/G]CCGGCCTGGAGGGCCGGATTGACCT
Phenotype
MIM: 172411
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PLA2G2A PubMed Links

Gene Details

Gene
PLA2G2A
Gene Name
phospholipase A2 group IIA
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000300.3 Intron NP_000291.1
NM_001161727.1 Intron NP_001155199.1
NM_001161728.1 Intron NP_001155200.1
NM_001161729.1 Intron NP_001155201.1

View Full Product Details