Product Details

SNP ID
rs11669824
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:58186902 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTGCAGTAGGATAGCATTTTCTC[C/G]TGAACACAGACCCCCACAAGCCCTG
Phenotype
MIM: 605467
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ZNF274 PubMed Links

Gene Details

Gene
ZNF274
Gene Name
zinc finger protein 274
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278734.1 Intron NP_001265663.1
NM_016324.3 Intron NP_057408.2
NM_016325.3 Intron NP_057409.1
NM_133502.2 Intron NP_598009.1
XM_011526327.1 Intron XP_011524629.1
XM_011526328.2 Intron XP_011524630.1
XM_017026174.1 Intron XP_016881663.1
XM_017026175.1 Intron XP_016881664.1

View Full Product Details