Product Details

SNP ID
rs7250137
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:6783547 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGAGTGCAATGGCGCGATCTCTGT[G/T]CAGTGCATCACGGCAACCTCCGCCT
Phenotype
MIM: 164875
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
VAV1 PubMed Links

Gene Details

Gene
VAV1
Gene Name
vav guanine nucleotide exchange factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001258206.1 Intron NP_001245135.1
NM_001258207.1 Intron NP_001245136.1
NM_005428.3 Intron NP_005419.2
XM_005259642.1 Intron XP_005259699.1

View Full Product Details