Product Details

SNP ID
rs3135960
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:34982914 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTTTGATTAGTCATCATGGACTAT[A/G]TAAGACGCTGGCCTTTGGAAAATTG
Phenotype
MIM: 610730 MIM: 600940
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCT6B PubMed Links

Gene Details

Gene
CCT6B
Gene Name
chaperonin containing TCP1 subunit 6B
There are no transcripts associated with this gene.

Gene
LIG3
Gene Name
DNA ligase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002311.4 Intron NP_002302.2
NM_013975.3 Intron NP_039269.2
XM_005257970.3 Intron XP_005258027.1
XM_006721896.3 Intron XP_006721959.1
XM_017024624.1 Intron XP_016880113.1

View Full Product Details