Product Details
- SNP ID
-
rs12293073
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:581279 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTTAGGGCTCACCCTTCACGTGCTG[C/T]GGCGGTGGATGTGACACTGGGACAC
- Phenotype
-
MIM: 611780
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LOC143666
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs111406154] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LOC143666
- Gene Name
- uncharacterized LOC143666
There are no transcripts associated with this gene.
- Gene
- PHRF1
- Gene Name
- PHD and ring finger domains 1
View Full Product Details