Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001144825.1 | Intron | NP_001138297.1 | |||
NM_001144826.1 | Intron | NP_001138298.1 | |||
NM_006695.4 | Intron | NP_006686.1 | |||
XM_005256964.3 | Intron | XP_005257021.1 | |||
XM_017024035.1 | Intron | XP_016879524.1 | |||
XM_017024036.1 | Intron | XP_016879525.1 | |||
XM_017024037.1 | Intron | XP_016879526.1 | |||
XM_017024038.1 | Intron | XP_016879527.1 | |||
XM_017024039.1 | Intron | XP_016879528.1 | |||
XM_017024040.1 | Intron | XP_016879529.1 |