Product Details

SNP ID
rs11868035
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:17811787 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATCCTCCCGTGCCACCCAGGGACCT[A/G]ATGGGGACAGAGCTGGGAGGTGAGA
Phenotype
MIM: 613486 MIM: 607642 MIM: 184756
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR33B PubMed Links
Additional Information
For this assay, SNP(s) [rs73981075] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
MIR33B
Gene Name
microRNA 33b
There are no transcripts associated with this gene.

Gene
MIR6777
Gene Name
microRNA 6777
There are no transcripts associated with this gene.

Gene
RAI1
Gene Name
retinoic acid induced 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030665.3 4473 Intron NP_109590.3
XM_017024025.1 4473 Intron XP_016879514.1
XM_017024026.1 4473 Intron XP_016879515.1
XM_017024027.1 4473 Intron XP_016879516.1
XM_017024028.1 4473 Intron XP_016879517.1
Gene
SREBF1
Gene Name
sterol regulatory element binding transcription factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005291.2 4473 UTR 3 NP_001005291.1
NM_001321096.2 4473 Intron NP_001308025.1
NM_004176.4 4473 UTR 3 NP_004167.3
XM_005256772.4 4473 Intron XP_005256829.1
XM_017024970.1 4473 Intron XP_016880459.1
XM_017024971.1 4473 Intron XP_016880460.1

View Full Product Details