Product Details

SNP ID
rs12052474
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:88700323 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCAGAGATGGCCCGGTAGTCTCTCT[C/T]CTCAAGGAATATTACTCCTATTTAT
Phenotype
MIM: 180430
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RPIA PubMed Links

Gene Details

Gene
RPIA
Gene Name
ribose 5-phosphate isomerase A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144563.2 Intron NP_653164.2

View Full Product Details