Product Details

SNP ID
rs11465341
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:75770987 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAAAGGAGGATAGAAAATACAAAAC[C/T]CGGTGATGCAAAAGCTGCATAAATT
Phenotype
MIM: 604697
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCL26 PubMed Links

Gene Details

Gene
CCL26
Gene Name
C-C motif chemokine ligand 26
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006072.4 Intron NP_006063.1
XM_017011671.1 Intron XP_016867160.1
XM_017011672.1 Intron XP_016867161.1

View Full Product Details