Product Details

SNP ID
rs11867471
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:47472068 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGAGCCTAGGAGTTCCAGCCTGTA[A/G]CATGCCATGATTGTGCCTGTGAATA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MRPL45P2 PubMed Links

Gene Details

Gene
MRPL45P2
Gene Name
mitochondrial ribosomal protein L45 pseudogene 2
There are no transcripts associated with this gene.

View Full Product Details