Product Details

SNP ID
rs11577731
Assay Type
Functionally tested
NCBI dbSNP Submissions
15
Location
Chr.1:145750494 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGAGGGCCTGTGTTTTCCAGTTGTC[T/C]TAAAAGCTACAAAAAAAGAGAAAGA
Phenotype
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
RNF115 PubMed Links

Gene Details

Gene
RNF115
Gene Name
ring finger protein 115
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014455.3 830 Missense Mutation AGA,GGA R194G NP_055270.1
XM_005272952.4 830 Missense Mutation AGA,GGA R161G XP_005273009.1

View Full Product Details