Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001197325.1 | Intron | NP_001184254.1 | |||
NM_001286035.1 | Intron | NP_001272964.1 | |||
NM_001286036.1 | Intron | NP_001272965.1 | |||
NM_001668.3 | Intron | NP_001659.1 | |||
NM_178427.2 | Intron | NP_848514.1 | |||
XM_005245151.1 | Intron | XP_005245208.1 | |||
XM_005245153.1 | Intron | XP_005245210.1 | |||
XM_005245154.3 | Intron | XP_005245211.1 | |||
XM_005245157.1 | Intron | XP_005245214.1 | |||
XM_011509542.2 | Intron | XP_011507844.1 | |||
XM_011509543.2 | Intron | XP_011507845.1 | |||
XM_011509544.2 | Intron | XP_011507846.1 | |||
XM_011509545.2 | Intron | XP_011507847.1 | |||
XM_011509546.1 | Intron | XP_011507848.1 | |||
XM_011509547.2 | Intron | XP_011507849.1 | |||
XM_017001288.1 | Intron | XP_016856777.1 | |||
XM_017001289.1 | Intron | XP_016856778.1 | |||
XM_017001290.1 | Intron | XP_016856779.1 | |||
XM_017001291.1 | Intron | XP_016856780.1 | |||
XM_017001292.1 | Intron | XP_016856781.1 | |||
XM_017001293.1 | Intron | XP_016856782.1 | |||
XM_017001294.1 | Intron | XP_016856783.1 | |||
XM_017001295.1 | Intron | XP_016856784.1 | |||
XM_017001296.1 | Intron | XP_016856785.1 |