Product Details

SNP ID
rs10305749
Assay Type
Functionally tested
NCBI dbSNP Submissions
18
Location
Chr.1:150812015 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCCCCCACCCCTTATCCTCACCCCA[A/G]TAGTTCTATTCTGAAAAGGGGGGAA
Phenotype
MIM: 126110 MIM: 601105
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARNT PubMed Links

Gene Details

Gene
ARNT
Gene Name
aryl hydrocarbon receptor nuclear translocator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001197325.1 2407 UTR 3 NP_001184254.1
NM_001286035.1 2407 UTR 3 NP_001272964.1
NM_001286036.1 2407 UTR 3 NP_001272965.1
NM_001668.3 2407 UTR 3 NP_001659.1
NM_178427.2 2407 UTR 3 NP_848514.1
XM_005245151.1 2407 UTR 3 XP_005245208.1
XM_005245153.1 2407 UTR 3 XP_005245210.1
XM_005245154.3 2407 UTR 3 XP_005245211.1
XM_005245157.1 2407 UTR 3 XP_005245214.1
XM_011509542.2 2407 UTR 3 XP_011507844.1
XM_011509543.2 2407 UTR 3 XP_011507845.1
XM_011509544.2 2407 UTR 3 XP_011507846.1
XM_011509545.2 2407 UTR 3 XP_011507847.1
XM_011509546.1 2407 UTR 3 XP_011507848.1
XM_011509547.2 2407 UTR 3 XP_011507849.1
XM_017001288.1 2407 UTR 3 XP_016856777.1
XM_017001289.1 2407 UTR 3 XP_016856778.1
XM_017001290.1 2407 UTR 3 XP_016856779.1
XM_017001291.1 2407 UTR 3 XP_016856780.1
XM_017001292.1 2407 UTR 3 XP_016856781.1
XM_017001293.1 2407 UTR 3 XP_016856782.1
XM_017001294.1 2407 UTR 3 XP_016856783.1
XM_017001295.1 2407 UTR 3 XP_016856784.1
XM_017001296.1 2407 UTR 3 XP_016856785.1
Gene
CTSK
Gene Name
cathepsin K
There are no transcripts associated with this gene.

View Full Product Details