Product Details

SNP ID
rs12032259
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:150622710 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGGACACCAACAGGAAAGGGCTT[C/T]TGCCTCTCCAGCCCACACCCGAGCC
Phenotype
MIM: 603061
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ENSA PubMed Links

Gene Details

Gene
ENSA
Gene Name
endosulfine alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004436.2 516 UTR 3 NP_004427.1
NM_207042.1 516 UTR 3 NP_996925.1
NM_207043.1 516 Intron NP_996926.1
NM_207044.1 516 Intron NP_996927.1
NM_207045.1 516 UTR 3 NP_996928.1
NM_207046.1 516 UTR 3 NP_996929.1
NM_207047.1 516 Intron NP_996930.1
NM_207168.1 516 Intron NP_997051.1

View Full Product Details