Product Details

SNP ID
rs11102471
Assay Type
Functionally tested
NCBI dbSNP Submissions
37
Location
Chr.1:112397162 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTGGGCAGGAATGCAGTATATGTT[C/G]CTTGACAGTCAATGAATAACACCAT
Phenotype
MIM: 615100
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CTTNBP2NL PubMed Links

Gene Details

Gene
CTTNBP2NL
Gene Name
CTTNBP2 N-terminal like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018704.2 Intron NP_061174.1
XM_011541781.2 Intron XP_011540083.1
XM_017001806.1 Intron XP_016857295.1

View Full Product Details