Product Details

SNP ID
rs11102480
Assay Type
Functionally tested
NCBI dbSNP Submissions
15
Location
Chr.1:112481415 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGAAGAAACTTACCCTAAAATAAT[A/C]AAGGCCATTATGAAAAGTCCACAGC
Phenotype
MIM: 601968
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
WNT2B PubMed Links

Gene Details

Gene
WNT2B
Gene Name
Wnt family member 2B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291880.1 Intron NP_001278809.1
NM_004185.4 Intron NP_004176.2
NM_024494.2 Intron NP_078613.1

View Full Product Details