Product Details

SNP ID
rs12069411
Assay Type
Functionally tested
NCBI dbSNP Submissions
15
Location
Chr.1:113085930 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATTCTTGTGTTTAGTTCAGTTTAC[G/T]CCAAGAATATTCAAGATCATAAGCA
Phenotype
MIM: 608869
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LRIG2 PubMed Links

Gene Details

Gene
LRIG2
Gene Name
leucine rich repeats and immunoglobulin like domains 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001312686.1 Intron NP_001299615.1
NM_014813.2 Intron NP_055628.1
XM_005271369.2 Intron XP_005271426.1
XM_017002952.1 Intron XP_016858441.1

View Full Product Details