Product Details

SNP ID
rs6537789
Assay Type
Functionally tested
NCBI dbSNP Submissions
52
Location
Chr.1:113399486 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTAATCTCTCTTACTTGGAGGCCAG[A/C]AGTAGTACACAGTTGTCATAAAACT
Phenotype
MIM: 615943
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
MAGI3 PubMed Links

Gene Details

Gene
MAGI3
Gene Name
membrane associated guanylate kinase, WW and PDZ domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142782.1 Intron NP_001136254.1
NM_152900.2 Intron NP_690864.2
XM_005270737.3 Intron XP_005270794.1
XM_011541208.2 Intron XP_011539510.1
XM_017000974.1 Intron XP_016856463.1
XM_017000975.1 Intron XP_016856464.1

View Full Product Details