Product Details

SNP ID
hCV31476314
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.1:113077480 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGTCTTGGATCAGTATTGGGAAC[C/T]AGTGCTGTACTTGATTAATTTATTG
Phenotype
MIM: 608869
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC100996251 PubMed Links

Gene Details

Gene
LOC100996251
Gene Name
uncharacterized LOC100996251
There are no transcripts associated with this gene.

Gene
LRIG2
Gene Name
leucine rich repeats and immunoglobulin like domains 2
There are no transcripts associated with this gene.

View Full Product Details