Product Details

SNP ID
rs11588393
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:114772908 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTATTACTGGGATCAGATGCTTTTA[A/G]GACATCTTTAAAATATTTTGTTATT
Phenotype
MIM: 611656
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SIKE1 PubMed Links

Gene Details

Gene
SIKE1
Gene Name
suppressor of IKBKE 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001102396.1 2067 UTR 3 NP_001095866.1
NM_025073.2 2067 UTR 3 NP_079349.2

View Full Product Details