Product Details

SNP ID
rs12064849
Assay Type
Functionally tested
NCBI dbSNP Submissions
8
Location
Chr.1:114705886 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAACATACTTGGAAAACTCTAAGTA[C/T]ATACATACAAGGGCTATTGGCACAA
Phenotype
MIM: 164790
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NRAS PubMed Links

Gene Details

Gene
NRAS
Gene Name
neuroblastoma RAS viral oncogene homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002524.4 3032 UTR 3 NP_002515.1

View Full Product Details