Product Details

SNP ID
rs12609699
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:39344075 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTCTTGAAATCCCTCCAAGACCTT[C/G]CTGACACCACCAGAACCCCTTGAAT
Phenotype
MIM: 604104
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GMFG PubMed Links

Gene Details

Gene
GMFG
Gene Name
glia maturation factor gamma
There are no transcripts associated with this gene.

Gene
SAMD4B
Gene Name
sterile alpha motif domain containing 4B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303614.1 1291 Intron NP_001290543.1
NM_018028.3 1291 Intron NP_060498.2
XM_011527057.2 1291 Intron XP_011525359.1
XM_011527060.2 1291 Intron XP_011525362.1
XM_011527061.2 1291 Intron XP_011525363.1
XM_011527062.2 1291 Intron XP_011525364.1
XM_011527063.2 1291 Intron XP_011525365.1
XM_011527064.2 1291 Intron XP_011525366.1
XM_017026918.1 1291 Intron XP_016882407.1
XM_017026919.1 1291 Intron XP_016882408.1
XM_017026920.1 1291 Intron XP_016882409.1
XM_017026921.1 1291 Intron XP_016882410.1
XM_017026922.1 1291 Intron XP_016882411.1
XM_017026923.1 1291 UTR 5 XP_016882412.1

View Full Product Details