Product Details

SNP ID
rs7030554
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:81986523 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGTAAATATATGTATCATACATCCC[A/G]TGTTCACTTTGGAATGGGGTTTTAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SPATA31D1 PubMed Links

Gene Details

Gene
SPATA31D1
Gene Name
SPATA31 subfamily D member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001670.2 Intron NP_001001670.1
XM_017014710.1 Intron XP_016870199.1

View Full Product Details