Product Details

SNP ID
rs6803711
Assay Type
Functionally Tested
NCBI dbSNP Submissions
8
Location
Chr.3:64517667 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAGCCTGAAATCCCCCCACCTGAGA[C/T]AACTGCTCTTCATATATTGGTATTT
Phenotype
MIM: 605421
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
ADAMTS9 PubMed Links

Gene Details

Gene
ADAMTS9
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318781.1 Intron NP_001305710.1
NM_182920.1 Intron NP_891550.1

View Full Product Details