Product Details

SNP ID
rs12454232
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:79397017 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCAAGTTTGAAAAGGAACCGCCAA[G/T]TTTCGAGCTGCTTCTTTATAGTGAA
Phenotype
MIM: 600489
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
NFATC1 PubMed Links

Gene Details

Gene
NFATC1
Gene Name
nuclear factor of activated T-cells 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278669.1 Intron NP_001265598.1
NM_001278670.1 Intron NP_001265599.1
NM_001278672.1 Intron NP_001265601.1
NM_001278673.1 Intron NP_001265602.1
NM_001278675.1 Intron NP_001265604.1
NM_006162.4 Intron NP_006153.2
NM_172387.2 Intron NP_765975.1
NM_172388.2 Intron NP_765976.1
NM_172389.2 Intron NP_765977.1
NM_172390.2 Intron NP_765978.1
XM_017025783.1 Intron XP_016881272.1

View Full Product Details