Product Details

SNP ID
rs11883235
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:56595473 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGGTGCTGGTCCAGGGCGTGCGGGC[C/G]CAGGAGAGGGCGTGCAGGCCGGGAC
Phenotype
MIM: 194545
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ZNF71 PubMed Links

Gene Details

Gene
ZNF71
Gene Name
zinc finger protein 71
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021216.4 Intron NP_067039.1
XM_011527193.2 Intron XP_011525495.1
XM_017027078.1 Intron XP_016882567.1
XM_017027079.1 Intron XP_016882568.1
XM_017027080.1 Intron XP_016882569.1

View Full Product Details