Product Details
- SNP ID
-
rs12045100
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
11
- Location
-
Chr.1:101879240 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ATTTTACTAAACACTACATTTTTTT[C/T]TTTTGATGTCTGTATGGCATTATTT
- Phenotype
-
MIM: 607567
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
DNAJA1P5
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs199951196] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- DNAJA1P5
- Gene Name
- DnaJ heat shock protein family (Hsp40) member A1 pseudogene 5
There are no transcripts associated with this gene.
- Gene
- OLFM3
- Gene Name
- olfactomedin 3
View Full Product Details