Product Details

SNP ID
rs12064882
Assay Type
Functionally tested
NCBI dbSNP Submissions
15
Location
Chr.1:89107687 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACAAAGACCACTTGGCAGGAGTGA[C/T]GCTCTACTGTGCCCCATGGTTTGTT
Phenotype
MIM: 600412
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GBP2 PubMed Links

Gene Details

Gene
GBP2
Gene Name
guanylate binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004120.4 2578 UTR 3 NP_004111.2

View Full Product Details