Product Details

SNP ID
rs12086533
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:84512506 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATTGAGCAATCTTTGTATGCCAT[A/G]TTATCAAATCAGACAGGAAATAAAA
Phenotype
MIM: 600874
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GNG5 PubMed Links

Gene Details

Gene
GNG5
Gene Name
G protein subunit gamma 5
There are no transcripts associated with this gene.

Gene
SPATA1
Gene Name
spermatogenesis associated 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001310156.1 Intron NP_001297085.1
XM_011542514.2 Intron XP_011540816.1
XM_011542515.2 Intron XP_011540817.1
XM_011542516.1 Intron XP_011540818.1
XM_011542517.1 Intron XP_011540819.1
XM_011542519.2 Intron XP_011540821.1
XM_011542520.2 Intron XP_011540822.1
XM_011542521.2 Intron XP_011540823.1
XM_011542522.2 Intron XP_011540824.1

View Full Product Details