Product Details

SNP ID
rs12794229
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:62878214 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGACTTCAGCCATTTGGAGCCATGA[C/G]GATCTGTGAGTGAGATACTGTGTCA
Phenotype
MIM: 158070
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SLC3A2 PubMed Links

Gene Details

Gene
SLC3A2
Gene Name
solute carrier family 3 member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012662.2 Intron NP_001012680.1
NM_001012664.2 Intron NP_001012682.1
NM_001013251.2 Intron NP_001013269.1
NM_002394.5 Intron NP_002385.3

View Full Product Details