Product Details

SNP ID
rs10896167
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:67298418 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGCCTTGAAGAGTGTTCCGTGTGCG[C/T]GTGAGAAGCGTGTGTGTGGTTGTTA
Phenotype
MIM: 615126 MIM: 606780
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ANKRD13D PubMed Links

Gene Details

Gene
ANKRD13D
Gene Name
ankyrin repeat domain 13D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207354.2 Intron NP_997237.2
XM_011544985.1 Intron XP_011543287.1
XM_011544986.1 Intron XP_011543288.1
XM_011544987.2 Intron XP_011543289.1
XM_011544988.2 Intron XP_011543290.1
Gene
SSH3
Gene Name
slingshot protein phosphatase 3
There are no transcripts associated with this gene.

View Full Product Details