Product Details

SNP ID
rs7270738
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:38035741 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGGTGGGACCTCAAGAGGCAGTTA[A/C]AGTCCATCTCAGGTGTCATTTTTTT
Phenotype
MIM: 614694 MIM: 614425
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
RPRD1B PubMed Links

Gene Details

Gene
RPRD1B
Gene Name
regulation of nuclear pre-mRNA domain containing 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021215.3 Intron NP_067038.1
XM_005260477.3 Intron XP_005260534.1
XM_005260480.4 Intron XP_005260537.1
XM_011528954.2 Intron XP_011527256.1
XM_017027990.1 Intron XP_016883479.1
XM_017027991.1 Intron XP_016883480.1
Gene
TTI1
Gene Name
TELO2 interacting protein 1
There are no transcripts associated with this gene.

View Full Product Details