Product Details

SNP ID
rs13315672
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:102390172 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCTCCAAGGCACCTCAAATAAAC[A/G]TACAATCTATATTAAAACCATGTGC
Phenotype
MIM: 615915
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZPLD1 PubMed Links

Gene Details

Gene
ZPLD1
Gene Name
zona pellucida like domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175056.1 Intron NP_778226.1
XM_017005703.1 Intron XP_016861192.1
XM_017005704.1 Intron XP_016861193.1

View Full Product Details