Product Details

SNP ID
rs11643314
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:29993317 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCCAGGGTCCGTCGGTACAGCTCCC[C/A]TGGGGGTGCTGCTTCTCCTAAAGGG
Phenotype
MIM: 603365 MIM: 613199
Polymorphism
C/A, Transversion substitution
Allele Nomenclature
Literature Links
HIRIP3 PubMed Links

Gene Details

Gene
HIRIP3
Gene Name
HIRA interacting protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001197323.1 2052 UTR 3 NP_001184252.1
NM_003609.4 2052 Missense Mutation GGG,TGG G521W NP_003600.2
Gene
INO80E
Gene Name
INO80 complex subunit E
There are no transcripts associated with this gene.

Gene
TAOK2
Gene Name
TAO kinase 2
There are no transcripts associated with this gene.

View Full Product Details