Product Details
- SNP ID
-
rs12897819
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:94129360 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGGAGAAAGAGGAGAGAGAAGGGCA[C/G]AAAGGGAGGGAGGGGGAGGGAAGCA
- Phenotype
-
MIM: 611319
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
IFI27L2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs77127154] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- IFI27L2
- Gene Name
- interferon alpha inducible protein 27 like 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_032036.2 |
|
Intron |
|
|
NP_114425.1 |
View Full Product Details