Product Details

SNP ID
rs12447435
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:46479164 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTAGACCAATGTTTTCTAATTGTT[C/G]TGCTATCTCCTGCCTTGCTCTTTCT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ANKRD26P1 PubMed Links

Gene Details

Gene
ANKRD26P1
Gene Name
ankyrin repeat domain 26 pseudogene 1
There are no transcripts associated with this gene.

View Full Product Details