Product Details

SNP ID
rs7187430
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:66581709 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTTATTCCCAACACAGAGGAATA[A/G]GAAAACATGTTCATCTCTGCCTGAG
Phenotype
MIM: 607884 MIM: 607885
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CKLF-CMTM1 PubMed Links

Gene Details

Gene
CKLF-CMTM1
Gene Name
CKLF-CMTM1 readthrough
There are no transcripts associated with this gene.

Gene
CMTM1
Gene Name
CKLF like MARVEL transmembrane domain containing 1
There are no transcripts associated with this gene.

Gene
CMTM2
Gene Name
CKLF like MARVEL transmembrane domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199317.1 Intron NP_001186246.1
NM_144673.2 Intron NP_653274.1

View Full Product Details