Product Details

SNP ID
rs11645068
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:12664962 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCTCATCTAGACTGTAGTATCGG[G/T]GAACAATTTTCTCGGCGGTGACCAC
Phenotype
MIM: 615603
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CPPED1 PubMed Links

Gene Details

Gene
CPPED1
Gene Name
calcineurin like phosphoesterase domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001099455.1 554 Missense Mutation CAC,CCC H148P NP_001092925.1
NM_018340.2 554 Missense Mutation CAC,CCC H290P NP_060810.2

View Full Product Details