Product Details

SNP ID
rs12254334
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:17142946 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTTTCAGGGAGGAGCAGGGAGAAA[G/T]AAATCTACACTCTCTTGTCAAGACC
Phenotype
MIM: 602478
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TRDMT1 PubMed Links

Gene Details

Gene
TRDMT1
Gene Name
tRNA aspartic acid methyltransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321006.1 7707 UTR 3 NP_001307935.1
NM_001321007.1 7707 UTR 3 NP_001307936.1
NM_004412.6 7707 UTR 3 NP_004403.1
XM_005252372.4 7707 Intron XP_005252429.1
XM_005252374.4 7707 Intron XP_005252431.1
XM_005252375.4 7707 UTR 3 XP_005252432.1
XM_005252378.3 7707 Intron XP_005252435.1
XM_006717388.3 7707 UTR 3 XP_006717451.1
XM_011519343.2 7707 UTR 3 XP_011517645.1
XM_011519344.2 7707 UTR 3 XP_011517646.1
XM_011519345.2 7707 UTR 3 XP_011517647.1
XM_017015800.1 7707 UTR 3 XP_016871289.1
XM_017015801.1 7707 Intron XP_016871290.1
XM_017015802.1 7707 Intron XP_016871291.1
XM_017015803.1 7707 Intron XP_016871292.1
XM_017015804.1 7707 Intron XP_016871293.1
XM_017015805.1 7707 Intron XP_016871294.1
XM_017015806.1 7707 UTR 3 XP_016871295.1
XM_017015807.1 7707 Intron XP_016871296.1
XM_017015808.1 7707 Intron XP_016871297.1
XM_017015809.1 7707 UTR 3 XP_016871298.1
XM_017015810.1 7707 Intron XP_016871299.1
XM_017015811.1 7707 UTR 3 XP_016871300.1
XM_017015812.1 7707 UTR 3 XP_016871301.1

View Full Product Details