Product Details
- SNP ID
-
rs12357352
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:42458692 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TATCTAGTAAAACCCTCAGGACCTA[A/G]TGGCTACAATCTGCGTTAATAGCAT
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CCNYL2
PubMed Links
Gene Details
- Gene
- CCNYL2
- Gene Name
- cyclin Y-like 2 (pseudogene)
There are no transcripts associated with this gene.
View Full Product Details