Product Details

SNP ID
rs12352181
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.9:125206581 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGTGCTAGCTTTGGCTTCCATCA[A/G]ATTGAGCTTTGGGGGTAATCTGGAA
Phenotype
MIM: 605962
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
LOC105376271 PubMed Links

Gene Details

Gene
LOC105376271
Gene Name
uncharacterized LOC105376271
There are no transcripts associated with this gene.

Gene
RABEPK
Gene Name
Rab9 effector protein with kelch motifs
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001174152.1 Intron NP_001167623.1
NM_001174153.1 Intron NP_001167624.1
NM_005833.3 Intron NP_005824.2
XM_005251640.4 Intron XP_005251697.1
XM_005251641.4 Intron XP_005251698.1
XM_005251642.4 Intron XP_005251699.1
XM_005251644.4 Intron XP_005251701.1
XM_011518120.2 Intron XP_011516422.1
XM_017014177.1 Intron XP_016869666.1
XM_017014178.1 Intron XP_016869667.1
XM_017014179.1 Intron XP_016869668.1
XM_017014180.1 Intron XP_016869669.1
XM_017014181.1 Intron XP_016869670.1
XM_017014182.1 Intron XP_016869671.1
XM_017014183.1 Intron XP_016869672.1

View Full Product Details