Product Details

SNP ID
rs12602960
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:5511006 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCACCTAGTCAAACAACCGTTTGAC[G/T]CAAGCTTTGCTATTCCCTTCTTACA
Phenotype
MIM: 606636
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
NLRP1 PubMed Links

Gene Details

Gene
NLRP1
Gene Name
NLR family pyrin domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033053.2 Intron NP_001028225.1
NM_014922.4 Intron NP_055737.1
NM_033004.3 Intron NP_127497.1
NM_033006.3 Intron NP_127499.1
NM_033007.3 Intron NP_127500.1

View Full Product Details