Product Details

SNP ID
rs10753193
Assay Type
Functionally tested
NCBI dbSNP Submissions
77
Location
Chr.1:179365849 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTGACTTAGAACATAAGAAACTCC[A/G]AGAGGGTGTAGGGATTGTCTTAGCT
Phenotype
MIM: 102642
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AXDND1 PubMed Links

Gene Details

Gene
AXDND1
Gene Name
axonemal dynein light chain domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144696.5 109 UTR 5 NP_653297.3
XM_011509165.1 109 UTR 5 XP_011507467.1
XM_011509166.2 109 UTR 5 XP_011507468.1
XM_011509167.2 109 UTR 5 XP_011507469.1
XM_011509168.1 109 UTR 5 XP_011507470.1
XM_011509169.1 109 UTR 5 XP_011507471.1
XM_011509170.1 109 UTR 5 XP_011507472.1
XM_011509171.1 109 UTR 5 XP_011507473.1
XM_011509174.1 109 UTR 5 XP_011507476.1
XM_011509175.1 109 UTR 5 XP_011507477.1
XM_011509176.1 109 UTR 5 XP_011507478.1
XM_011509178.2 109 UTR 5 XP_011507480.1
XM_011509179.2 109 Intron XP_011507481.1
XM_011509180.1 109 UTR 5 XP_011507482.1
XM_011509181.2 109 Intron XP_011507483.1
XM_017000257.1 109 Intron XP_016855746.1
XM_017000258.1 109 Intron XP_016855747.1
XM_017000259.1 109 Intron XP_016855748.1
Gene
SOAT1
Gene Name
sterol O-acyltransferase 1
There are no transcripts associated with this gene.

View Full Product Details