Product Details

SNP ID
rs11571123
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:204155345 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATGGCCATTTTGCCCCATCGTGGA[C/G]TGCACCACTCCTATGTTTAGCCCCT
Phenotype
MIM: 609859 MIM: 179820
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ETNK2 PubMed Links

Gene Details

Gene
ETNK2
Gene Name
ethanolamine kinase 2
There are no transcripts associated with this gene.

Gene
REN
Gene Name
renin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000537.3 Intron NP_000528.1

View Full Product Details